Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Polypropylene glycol

Catalog No. 813501L Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
81-350-1L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 81-350-1L Supplier Sigma Aldrich Supplier No. 813501L
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Propylene glycol is an aliphatic alcohol. It is an addition polymer of propylene glycol and water represented as H[OCH3]nOH in which n represents the average number of oxypropylene groups.Propylene glycol (PPG) may be used as a viscosity decreasing agent, a solvent and a fragrance ingredient in cosmetics. PPG may be used as a good swelling agent for the synthesis of large pore mesoporous materials.

Specifications

CAS 25322-69-4
Density 1.01 g/mL (at 20°C)
Refractive Index n20/D 1.447
Quantity 1 L
MDL Number MFCD00084446
Molecular Weight (g/mol) 446
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.