Learn More
Medchemexpress LLC Polypropylene glycol 2000 | 25322-69-4 | MFCD00084446 | 2,000 g/mol (Mn) | HO(C3H6O)nC3H6OH | 100 G
SDP
Catalog No. 5000283370
Encompass_Preferred
Polypropylene glycol 2000 is a polyether polymer (average Mn ≈ 2,000 g/mol) supplied as a research-grade reagent for laboratory formulation and analytical use. It offers good solubility in many organic systems and is commonly used as a lubricant, solubilizer, and antifoaming agent in research formulations. Intended for research use only, not for therapeutic or human use.
  • Average molecular weight ~2,000 g/mol (Mn).
  • Functions as a lubricant, solubilizer, and antifoaming agent.
  • Suitable for formulation and analytical research applications.
  • Provided in research pack sizes (for example, 100 G).
  • CAS number 25322-69-4 for unambiguous identification.
  • For research use only; not for human or therapeutic use.
Catalog No. 50-002-83370 Supplier Medchemexpress LLC Supplier No. HYW133920100G
Polypropylene glycol 2000 is a polyether polymer (average Mn ≈ 2,000 g/mol) supplied as a research-grade reagent for laboratory formulation and analytical use. It offers good solubility in many organic systems and is commonly used as a lubricant, solubilizer, and antifoaming agent in research formulations. Intended for research use only, not for therapeutic or human use.
  • Average molecular weight ~2,000 g/mol (Mn).
  • Functions as a lubricant, solubilizer, and antifoaming agent.
  • Suitable for formulation and analytical research applications.
  • Provided in research pack sizes (for example, 100 G).
  • CAS number 25322-69-4 for unambiguous identification.
  • For research use only; not for human or therapeutic use.