Learn More
Medchemexpress LLC Polypropylene glycol | 25322-69-4 | MFCD00084446 | 2000.00 | (C3H6O)n | 500 G
SDP
Catalog No. 5000283127
Encompass_Preferred
Polypropylene glycol 2000 is a high-molecular-weight polyether liquid used in laboratory and biochemical applications as a solvent, dispersant, and reagent modifier. It appears as a colorless to light-yellow viscous liquid with an average molecular weight of about 2000, provided with supporting documentation for safe laboratory use.
  • High average molecular weight (~2000) for consistent performance.
  • Colorless to light-yellow liquid suitable for solvents and formulations.
  • Useful as a dispersant, reagent modifier, and solvent in assays and preparations.
  • Analytical documentation available, including SDS, COA, and data sheet.
  • Low acid value and defined hydroxyl value for reproducible chemistry.
  • Available in laboratory pack sizes for routine use.
Catalog No. 50-002-83127 Supplier Medchemexpress LLC Supplier No. HYW133920500G
Polypropylene glycol 2000 is a high-molecular-weight polyether liquid used in laboratory and biochemical applications as a solvent, dispersant, and reagent modifier. It appears as a colorless to light-yellow viscous liquid with an average molecular weight of about 2000, provided with supporting documentation for safe laboratory use.
  • High average molecular weight (~2000) for consistent performance.
  • Colorless to light-yellow liquid suitable for solvents and formulations.
  • Useful as a dispersant, reagent modifier, and solvent in assays and preparations.
  • Analytical documentation available, including SDS, COA, and data sheet.
  • Low acid value and defined hydroxyl value for reproducible chemistry.
  • Available in laboratory pack sizes for routine use.