Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Polypropylene glycol | 25322-69-4 | MFCD00084446 | 2000.00 | C3nH6n+2On+1 | 1 KG
SDP

Catalog No. 5000283287
Encompass_Preferred

Polypropylene glycol 2000 is a liquid polymer (average molecular weight approximately 2000 Da) used as a solvent, excipient, and formulation component in research applications. It is typically colorless to light yellow and is supplied with storage and solubility guidance for laboratory use.

  • Liquid polymer with average molecular weight approximately 2000 Da.
  • Colorless to light yellow appearance.
  • Used as a solvent, excipient, and formulation agent in laboratories.
  • Soluble in DMSO; warming and ultrasonic treatment improve solubility.
  • Storage recommendations: long-term at -20°C, short-term at 4°C.
  • Available in kilogram-scale packaging for research needs.

Catalog No. 50-002-83287 Supplier Medchemexpress LLC Supplier No. HYW1339201KG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Polypropylene glycol 2000 is a liquid polymer (average molecular weight approximately 2000 Da) used as a solvent, excipient, and formulation component in research applications. It is typically colorless to light yellow and is supplied with storage and solubility guidance for laboratory use.

  • Liquid polymer with average molecular weight approximately 2000 Da.
  • Colorless to light yellow appearance.
  • Used as a solvent, excipient, and formulation agent in laboratories.
  • Soluble in DMSO; warming and ultrasonic treatment improve solubility.
  • Storage recommendations: long-term at -20°C, short-term at 4°C.
  • Available in kilogram-scale packaging for research needs.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.