Learn More
Andwin Scientific POLYPROPYLENE GRAD CYLINDER 1L
SDP
Catalog No. NC9418245
Encompass_Preferred
Polypropylene Graduated Cylinders 1000mL cylinders feature a single scale with raised numeral graduations in 10mL increments. Non-wetting polypropylene surface eliminates concave menisci. Flared top rim. Large spout. Hexagonal base. Chemically resista
Catalog No. NC9418245 Supplier Andwin Scientific Supplier No. 65000012
Polypropylene Graduated Cylinders 1000mL cylinders feature a single scale with raised numeral graduations in 10mL increments. Non-wetting polypropylene surface eliminates concave menisci. Flared top rim. Large spout. Hexagonal base. Chemically resista