Learn More
International Enviroguard™ Polypropylene Lab Coats with Three Pockets, Knit Wrist and Collar
Catalog No. 17183986 Shop All International Enviroguard Systems Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
4X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-183-986 Medium
17-183-987 Large
17-183-985 X-Large
17-183-990 2X-Large
17-183-988 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 17-183-986 Supplier International Enviroguard™ Supplier No. K2047DBM
Only null left
Add to Cart
Add to Cart
Dark blue lab coats are made of heavyweight polypropylene to provide affordable protection against dirt, grime, and particulates in non-hazardous environments.
  • Polypropylene lab coats are breathable to help keep users cool throughout their workday
  • Low-cost protection against dirt, grime, dust, and dry particulates
  • Three pockets allow wearers to take small items with them on the go, like eyeglasses or pens
  • Possible applications include food processing, pharmaceuticals, warehousing, general cleaning, environmental cleanup, and more
Specifications
Product Type General Purpose Lab Coat
Size Medium
Gender Unisex
Material Polypropylene
Color Navy
Closure Type Snaps
Cuff Style Knit
Disposable Single-use
Sterility Non-sterile
Resistance Particulates
Style Mid-length
No. of Pockets 3
Latex Free Yes
Model K2047DB
Autoclavable Not Autoclavable