Learn More
International Enviroguard™ Polypropylene Lab Coats with Three Pockets, Knit Wrist and Collar
Catalog No. 17183990 Shop All International Enviroguard Systems Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-183-990 2X-Large
17-183-986 Medium
17-183-987 Large
17-183-985 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 17-183-990 Supplier International Enviroguard™ Supplier No. K2047DB2XL
Add to Cart
Add to Cart
Dark blue lab coats are made of heavyweight polypropylene to provide affordable protection against dirt, grime, and particulates in non-hazardous environments.
  • Polypropylene lab coats are breathable to help keep users cool throughout their workday
  • Low-cost protection against dirt, grime, dust, and dry particulates
  • Three pockets allow wearers to take small items with them on the go, like eyeglasses or pens
  • Possible applications include food processing, pharmaceuticals, warehousing, general cleaning, environmental cleanup, and more
Specifications
Product Type General Purpose Lab Coat
Size 2X-Large
Gender Unisex
Material Polypropylene
Color Navy
Closure Type Snaps
Cuff Style Knit
Disposable Single-use
Sterility Non-sterile
Resistance Particulates
Style Mid-length
No. of Pockets 3
Latex Free Yes
Model K2047DB
Seam Type Serged
Autoclavable Not Autoclavable