Learn More
International Enviroguard™ Polypropylene Lab Coats, Three Pocket
Catalog No. 19472472 Shop All International Enviroguard Systems Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-472-472 Medium
19-472-473 Large
19-472-474 X-Large
19-472-475 2X-Large
19-472-476 3X-Large
19-472-477 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 19-472-472 Supplier International Enviroguard™ Supplier No. K2045M
Add to Cart
Add to Cart
Polypropylene lab coats feature three convenient pockets and provide economical, lightweight protection against dirt, grime, and particulates in non-hazardous environments.
  • Polypropylene lab coats protect from dust, dirt, grime, and other dry particulates in non-hazardous environments
  • Ideal for laboratories, pharmaceuticals, food processing, environmental cleanup sites, controlled environments
  • Breathable fabric helps prevent heat stress and keeps wearers cool over long hours
  • Three spacious pockets to carry eyeglasses, pens, and other small instruments
  • Low-cost protection
Specifications
Product Type Controlled Environments Lab Coat
Size Medium
Gender Unisex
Material Polypropylene
Color White
Closure Type Snaps
Cuff Style Knit
Disposable Single-use
Sterility Non-sterile
Resistance Particulates
No. of Pockets 3
Collar Type Standard
Seam Type Serged