Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tians Polypropylene Lab Coats

Catalog No. 19063546 Shop All Tians International Products
Change view
Click to view available options
Size:
Large
X-Large
2X-Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-063-546 X-Large
19-038-440 Large
19-038-442 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-063-546 Supplier Tians Supplier No. 845880XL
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Comfortable with a cloth-like feel

  • Breathable low-lint polypropylene fabric
  • Folded collar
  • Snap front closure
  • Regular style coat features a breast pocket and cut wrists
  • Cleanroom coat style features knit wrists
  • White

Specifications

Product Type Controlled Environments Lab Coat
Size X-Large
Material Polypropylene
Color White
Closure Type Snaps
Cuff Style Open
Disposable Single-use
No. of Pockets 1
For Use With (Application) For cleanroom use
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.