Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Keystone Safety Blue Polypropylene Lab Coats, Elastic Wrists
SDP

Catalog No. 19061450
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-061-450 4X-Large
19-061-445 Medium
19-061-448 2X-Large
19-061-444 Small
19-061-449 3X-Large
19-061-447 X-Large
19-061-446 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 19-061-450 Supplier Keystone Safety Supplier No. LC0BENW4XLBLUE
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Cool and comfortable polypropylene lab coats are ideal for nonhazardous areas where a low particulate barrier is needed.

  • Lab coats provide protection against dirt, grime, and non-hazardous dry particulates
  • Non-woven polypropylene material is durable and low linting
  • Ideal for use in food manufacturing, light industrial, and medical environments
  • Full collar, elastic wrists
  • 3-pocket style available
  • Packaged 30 per case
  • Meets ANSI 101-1996

Specifications

Product Type General Purpose Lab Coat
Size 4X-Large
Gender Unisex
Material Polypropylene
Color Blue
Closure Type Snaps
Cuff Style Elastic
Disposable Single-use
Sterility Non-sterile
No. of Pockets None
Collar Type Single
Latex Free Yes
Model LCO-BE-NW
Certifications/Compliance ANSI 101-1996
For Use With (Application) Food, Manufacturing, Light Industrial, Medical
Weight (English) 1.4 oz.
Weight (Metric) 40 g
Autoclavable Not Autoclavable
No. per Pack 30/Pk.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.