Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Duran™ Polypropylene Membrane Screw Caps
SDP

Catalog No. 09841266 Shop All DWK Life Sciences Products
Change view
Click to view available options
Thread Size (Metric):
GL 25
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Thread Size (Metric)
09-841-266 GL 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09-841-266 Supplier Duran™ Supplier No. 1183250

missing translation for 'validMainframeMsg'

Specifications

Color Blue
Material Polypropylene
Quantity 5 Pk.
Thread Size (Metric) GL 25
Height (Metric) 19 mm
For Use With (Equipment) Pressure Equalization
Diameter (Metric) 33 mm
Product Type Membrane Screw Cap
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.