Learn More
Sigma Aldrich Fine Chemicals Biosciences Polypropylene NMR tube rac1EA
Catalog No. 502899996
Encompass
Polypropylene rack is steam autoclavable at 121C (250f)Dimensions 21 cm x 11 cm x 22 cm H (8 3/8 in. x 4 1/2 in. x 8 3/4 in.)Rugged and made specifically to support and safely store Nuclear Magnetic Resonance tubesUpper deck has holes lower deck has hemispherical wells to protect tipHandle in end plates for easy transportTube size 3 or 5 mm O.D. x 178 mm longHoles 72
Catalog No. 50-289-9996 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z1182571EA
Polypropylene rack is steam autoclavable at 121C (250f)Dimensions 21 cm x 11 cm x 22 cm H (8 3/8 in. x 4 1/2 in. x 8 3/4 in.)Rugged and made specifically to support and safely store Nuclear Magnetic Resonance tubesUpper deck has holes lower deck has hemispherical wells to protect tipHandle in end plates for easy transportTube size 3 or 5 mm O.D. x 178 mm longHoles 72