Learn More
Alkali Scientific Polypropylene PFAS Free Vial, 12x32mm (Screw Neck), with Cap and PTFE/Silicone Septum, 300µL Volume, 100/Pk
SDP
Catalog No. 5000662395
Encompass_Preferred
Engineered for high performance PFAS Analysis, Liquid Chromatography (HPLC) and LC MS applications, our 12 x 32 mm polypropylene vials deliver exceptional chemical resistance, low analyte adsorption, and reliable compatibility with autosamplers. Each vial features a precision molded screw neck and comes preassembled with a 9 mm open top cap and a PTFE/silicone septum to ensure clean, leak resistant performance—even after multiple injections. Chemical resistant polypropylene construction — ideal for biological and pH sensitive samples 300 µL capacity — minimizes sample waste for limited volume applications PTFE/silicone septum — supports repeated autosampler injections without coring or vacuum formation Leak proof 9 mm screw cap — ensures a secure, airtight seal Compatible with most major HPLC and LC MS autosamplers — including Waters, Agilent, Shimadzu, and more Non glass design — eliminates concerns with analyte binding or glass reactivity Perfect for laboratories handling sensitive.
Catalog No. 50-006-62395 Supplier Alkali Scientific Supplier No. CV5000
Engineered for high performance PFAS Analysis, Liquid Chromatography (HPLC) and LC MS applications, our 12 x 32 mm polypropylene vials deliver exceptional chemical resistance, low analyte adsorption, and reliable compatibility with autosamplers. Each vial features a precision molded screw neck and comes preassembled with a 9 mm open top cap and a PTFE/silicone septum to ensure clean, leak resistant performance—even after multiple injections. Chemical resistant polypropylene construction — ideal for biological and pH sensitive samples 300 µL capacity — minimizes sample waste for limited volume applications PTFE/silicone septum — supports repeated autosampler injections without coring or vacuum formation Leak proof 9 mm screw cap — ensures a secure, airtight seal Compatible with most major HPLC and LC MS autosamplers — including Waters, Agilent, Shimadzu, and more Non glass design — eliminates concerns with analyte binding or glass reactivity Perfect for laboratories handling sensitive.