Learn More
Bellco Glass, Inc. CELLTREAT Pasteur Pipets offer the aspirating precision pf a glass pipet and the safety benefits of plastic. Polypropylene provides the chemical resistivity needed for many applications.Specifications Size 5.75in Pack Description 200 Bulk Packed Box Case Quantity 1000
Supplier: Bellco Glass, Inc. 229287000
CELLTREAT Pasteur Pipets offer the aspirating precision pf a glass pipet and the safety benefits of plastic. Polypropylene provides the chemical resistivity needed for many applications.Specifications Size 5.75in Pack Description 200 Bulk Packed Box Case Quantity 1000