Learn More
Bellco Glass, Inc. Polypropylene Plasteur Pipet, 5.75 Inch Length, Bulk Packed, Sterile
SDP
Catalog No. 502329884
Encompass_Preferred
DescriptionThe Plasteur Plastic Pasteur Pipet offers the aspirating precision of a glass pipet and the safety benefits of plasticFine tip is ideal for precision aspiratingPolypropylene provides the chemical resistivity needed for many applicationsNo more broken glass: more durable than standard glass pipetsFlexible plastic helps prevent glove or skin puncturesWill not shatter, eliminating losses due to broken glassNo damage during shipping and handling as compared to traditional glass pipetsEliminate “sharps” disposalCan be autoclaved PPP Low Temp Autoclave Guideline Pack Description25 Bulk Packed/BagCase Quantity500SterileYesAutoclavableYesPrimary ColorClearPrimary MaterialPolypropyleneGraduatedNo
Catalog No. 50-232-9884 Supplier Bellco Glass, Inc. Supplier No. 229276000
DescriptionThe Plasteur Plastic Pasteur Pipet offers the aspirating precision of a glass pipet and the safety benefits of plasticFine tip is ideal for precision aspiratingPolypropylene provides the chemical resistivity needed for many applicationsNo more broken glass: more durable than standard glass pipetsFlexible plastic helps prevent glove or skin puncturesWill not shatter, eliminating losses due to broken glassNo damage during shipping and handling as compared to traditional glass pipetsEliminate “sharps” disposalCan be autoclaved PPP Low Temp Autoclave Guideline Pack Description25 Bulk Packed/BagCase Quantity500SterileYesAutoclavableYesPrimary ColorClearPrimary MaterialPolypropyleneGraduatedNo