Learn More
Bellco Glass, Inc. CELLTREAT Pasteur Pipets offer the aspirating precision pf a glass pipet and the safety benefits of plastic. Polypropylene provides the chemical resistivity needed for many applications.Specifications Size 5.75in Pack Description 200 Bulk Packed Box Case Quantity 1000
SDP
Catalog No. 502329305
Encompass_Preferred
CELLTREAT Pasteur Pipets offer the aspirating precision pf a glass pipet and the safety benefits of plastic. Polypropylene provides the chemical resistivity needed for many applications.Specifications Size 5.75in Pack Description 200 Bulk Packed Box Case Quantity 1000
Catalog No. 50-232-9305 Supplier Bellco Glass, Inc. Supplier No. 229286000
CELLTREAT Pasteur Pipets offer the aspirating precision pf a glass pipet and the safety benefits of plastic. Polypropylene provides the chemical resistivity needed for many applications.Specifications Size 5.75in Pack Description 200 Bulk Packed Box Case Quantity 1000