Learn More
AirSea USA 330ml plastic Biotube with 51mm bung – a suitable inner packaging for the transportation of infectious and biological substances, Category A UN 2814 and UN 2900, Category B UN 3373.
Supplier:  AirSea USA 330
Encompass_Preferred
330ml plastic Biotube with 51mm bung – a suitable inner packaging for the transportation of infectious substances. Approved with Code 332 (Biopack-1 330ml Class 6.2 infectious and biological substance packaging) and Code 333 (Temperature Control Packaging, Class 6.2 Infectious and Biological Substances)
Catalog No. NC2101114