Learn More
Alkali Scientific ThermaSeal™ Polypropylene Sealing Film, for Classic PCR or ELISA plates, Adhesive, 100/cs
SDP
Catalog No. 502336213
Encompass_Preferred
This transparent sealing film consists of a 2.0 mil polyolefin film coated on one side with a pressure sensitive acrylate adhesive which does not interfere with cycle reactions. It is ideal for reducing well-to-well contamination and/or spill over in sensitive PCR applications where the minimization of evaporation and vapor loss is critical. ThermaSeal™ Sealing Film was developed with the assistance of a major cycler manufacturer for PCR applications. Not only does it offer low-autofluorescence but it will prevent vapor loss and is thermostable and functional from -70 °C to +95 °C. Certified RNAse, DNAse and DNA-free. DMSO resistant. Note: Performance may depend upon the specific collection/sample vessel used as well as the specific conditions to which it is subjected. 100 per pack.
Catalog No. 50-233-6213 Supplier Alkali Scientific Supplier No. SF2ML
This transparent sealing film consists of a 2.0 mil polyolefin film coated on one side with a pressure sensitive acrylate adhesive which does not interfere with cycle reactions. It is ideal for reducing well-to-well contamination and/or spill over in sensitive PCR applications where the minimization of evaporation and vapor loss is critical. ThermaSeal™ Sealing Film was developed with the assistance of a major cycler manufacturer for PCR applications. Not only does it offer low-autofluorescence but it will prevent vapor loss and is thermostable and functional from -70 °C to +95 °C. Certified RNAse, DNAse and DNA-free. DMSO resistant. Note: Performance may depend upon the specific collection/sample vessel used as well as the specific conditions to which it is subjected. 100 per pack.