Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Keystone Safety Polypropylene Shoe Covers
SDP

Catalog No. 19055171
Change view
Click to view available options
Size:
Large
Universal
Color:
Blue
White
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Color
19-055-171 Universal Blue
19-055-176 Large White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-055-171 Supplier Keystone Safety Supplier No. SCNWILRGBLUE
Add to Cart
Add to Cart
This item is not returnable. View return policy

Ideal for contamination control and safety

 

  • 150 pair/case
  • White or Blue 

 

Specifications

Size Universal
Material Polypropylene
Color Blue
For Use With (Application) Ideal for contamination control and safety
Packaging Quantity 150 Pairs/Cs.
Resistance Fluid
Product Type Controlled Environments Shoe Covers
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.