Learn More
Andwin Scientific POLYPROPYLENE SNAP CAPS
SDP
Catalog No. NC9003112
Encompass_Preferred
Made with polyethylene, these clear caps are used to securely cover 2 mL snap-cap vials and are ideal for routine analyses. The caps are economical and moderately resealble. This product is sold in a pack of 500.
Catalog No. NC9003112 Supplier Andwin Scientific Supplier No. HP51820550
Made with polyethylene, these clear caps are used to securely cover 2 mL snap-cap vials and are ideal for routine analyses. The caps are economical and moderately resealble. This product is sold in a pack of 500.