Learn More
Hillrom Polypropylene Speculum
Catalog No. 22355153
Change view
Click to view available options
Size (Metric):
5 mm
Catalog No. Size (Metric)
22-355-153 5 mm
1 options
This item is not returnable. View return policy
Catalog No. 22-355-153 Supplier Hillrom Supplier No. WEL 22005
Add to Cart
Add to Cart
This item is not returnable. View return policy
This Polypropylene Speculum is lightweight and durable. It can be autoclaved, boiled or cleaned with standard germicides.
  • The specula may be autoclaved, boiled, or sterilized in solution and can be attached easily
  • The material used in the Polypropylene Specula is lightweight but durable
  • For use with Pneumatic and Operating Otoscopes
  • Style: Round Tip
Specifications
Disposable Reusable
For Use With (Equipment) Pneumatic Otoscope, Operating Otoscope
Product Type Otoscope Specula
Size (Metric) 5 mm