Learn More
Kuhner Shaker Inc POLYPROPYLENE SQUARE DEEPWELL
Catalog No. 502570138
Encompass
Polypropylene square deepwell MTP (re-usable, 96 wells of 2.2ml volume, flattened, detoxified and with slots for pins)
Catalog No. 50-257-0138 Supplier Kuhner Shaker Inc Supplier No. SMCR1496A
Polypropylene square deepwell MTP (re-usable, 96 wells of 2.2ml volume, flattened, detoxified and with slots for pins)