Learn More
Justrite Polypropylene Strap Assembly with Buckle
Catalog No. 19085876 Shop All Justrite Safety Group Products
Change view
Click to view available options
Includes:
Nylon Buckle
Steel Buckle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
19-085-876 Steel Buckle
19-085-878 Nylon Buckle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-085-876 Supplier Justrite Supplier No. 35406
Add to Cart
Add to Cart
Heavy-duty polypropylene support straps for quick securement of gas cylinders.
  • For quick securement of gas cylinders
  • Straps made of heavy-duty polypropylene
  • 54 in. long
  • Choose between nylon or steel buckle
Specifications
Type Gas Cylinder Stand
Holds 1 Cylinder
Load Bearing Capacity (English) 1000 lb.
Load Bearing Capacity (Metric) 454 kg
Material Polypropylene Alloy
Includes Steel Buckle