Learn More
Justrite Polypropylene Strap Assembly with Buckle
Catalog No. 19085878 Shop All Justrite Safety Group Products
Change view
Click to view available options
Includes:
Nylon Buckle
Steel Buckle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
19-085-878 Nylon Buckle
19-085-876 Steel Buckle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-085-878 Supplier Justrite Supplier No. 35410
  • $17.25
    $14.65 / Each of 1
    Save $2.60
    15% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Heavy-duty polypropylene support straps for quick securement of gas cylinders.
  • For quick securement of gas cylinders
  • Straps made of heavy-duty polypropylene
  • 54 in. long
  • Choose between nylon or steel buckle
Specifications
Type Gas Cylinder Stand
Holds 1 Cylinder
Load Bearing Capacity (English) 1000 lb.
Load Bearing Capacity (Metric) 454 kg
Material Polypropylene Alloy
Includes Nylon Buckle