Learn More
US Plastic Corporation 12" L x 14" W x 3" Hgt. Polypropylene Tamco® Tray
SDP
Catalog No. NC2818737
Encompass
Welded Polypropylene constructionWall thickness: 1/4"Rigid, sturdy constructionCorrosion resistantDimensions are inside dimensionsMaximum temperature: 200°FMaterials meet FDA standards
Catalog No. NC2818737 Supplier US Plastic Corporation Supplier No. 014659
Welded Polypropylene constructionWall thickness: 1/4"Rigid, sturdy constructionCorrosion resistantDimensions are inside dimensionsMaximum temperature: 200°FMaterials meet FDA standards