Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WHEATON™ Polypropylene Vial With Insert
SDP

Catalog No. 01672742 Shop All DWK Life Sciences Products
Change view
Click to view available options
Autoclavable:
Autoclavable
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Autoclavable
01-672-742 Autoclavable
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-672-742 Supplier WHEATON™ Supplier No. W225181
Add to Cart
Add to Cart

Polypropylene vial made of chemical resistant polypropylene

  • Manufactured from chemical resistant polypropylene
  • Economical alternative to glass
  • Vials are packaged 100 per shelf pack and 10 packs per case
  • Conical interior ensures maximum retrieval of contents without the hassle of using removable inserts
TRUSTED_SUSTAINABILITY

Specifications

Color Natural
Diameter (Metric) Outer 12 mm
Material Polypropylene
Sterility Non-sterile
Volume (Metric) 300 μL
Quantity 1000/Pk.
Marking Spot No
Autoclavable Autoclavable
Height (Metric) 32 mm
Preservative None
Product Type Vial
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.