Learn More
Bio Rad Laboratories POLYPRP COLMN 2ML 9CM LNG 50PK
Catalog No. NC9257122
Encompass
Poly-Prep Columns, 50pk, Height 9cm, Hold up to 2mL of chromatography media and 10mL or eluent or sample in an integral reservoir, Includes end caps and tip closures
Catalog No. NC9257122 Supplier Bio Rad Laboratories Supplier No. 7311550
Poly-Prep Columns, 50pk, Height 9cm, Hold up to 2mL of chromatography media and 10mL or eluent or sample in an integral reservoir, Includes end caps and tip closures