Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Polypyrrole

Catalog No. 57703025G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
57-703-025G 25 g
57-703-05G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 57-703-025G Supplier Sigma Aldrich Supplier No. 57703025G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Polypyrrole (PPy) is a widely used highly conductive and highly stable heterocyclic polymer. It is majorly used in electrochemical applications. It is a cost-efficient conducting polymer that has a stable oxidation and high solubility in water. It exhibits a high degree of flexibility than polyaniline and has a density that facilitates a higher capacitance per unit volume.PPy may be prepared by a standard electrochemical technique. It may also be prepared by reacting -napthalene sulfonic acid (NSA) and ammonium peroxo-disulfate in aqueous medium. The charges on the surfaces can be easily modified by doping the polymer during its synthesis. Solubility and conductivity measurements of PPy doped with camphor sulfonic and dodecyl benzene sulfonic acid have been reported. Lectrosensitivity and lower oxidation potential of PPy make it potentially useful for drug delivery, chemical sensors, batteries, ion selective electrodes, biosensor and biochemistry research.PPy is mainly used in the fabrication of a variety of electrochemical devices, which include supercapacitors, chemical sensors, dye sensitized solar cells and lithium-ion batteries.Stable up to at least 290°C in air

Specifications

CAS 30604-81-0
MDL Number MFCD00284347
Quantity 25 g
Synonym PPy
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.