Learn More
Beckman Coulter POLYQII VIAL 18 ML 5 TRAYS/100
Catalog No. NC9277900
Encompass_Preferred
Poly-Q Translucent Polyethylene 18mL Twist-On Cap 27.3 x 59.5mm Scintillation Vial, Unbreakable, Leakproof, No Serrations, Ideal for Cocktails
Catalog No. NC9277900 Supplier Beckman Coulter Supplier No. 566350
Poly-Q Translucent Polyethylene 18mL Twist-On Cap 27.3 x 59.5mm Scintillation Vial, Unbreakable, Leakproof, No Serrations, Ideal for Cocktails