Learn More
AdipoGen PolyR-3-hydroxybutyric acid
SDP
Supplier:  AdipoGen CDXP0074G010
Encompass_Preferred
Chemical. CAS 29435-48-1. Formulan. MW 104.1. PHB is a fully biodegradable polyester with optical activity, piezoelectricity, and very good barrier properties. PHB is a partially crystalline material with a high melting temperature and a high degree of crystallinity. PHB is a thermoplastic, belonging to the family of polyhydroxyalkanoate PHAs. It has physical- and mechanical properties comparable to those of isotactic polypropylene iPP. Different bacterial types of microorganisms produce PHB from renewable sources from sugar and molasses as intracellular storage materials. It can be used in applications of drug-loaded medical devices and microspheres as implantable and injectable therapeutic systems in medicine for treatment of various diseases. Besides application of PHB for producing of medical devices and systems of sustained drug delivery, PHB can be used for production of systems for controlled release of activators or inhibitors of enzymes. The use of these sys
Catalog No. 50-256-0260