Learn More
Enzo Life Sciences Polysialic acid Recombinant monoclonal antibody (735) (Mouse IgG2a kappa) (200µg)
SDP
Supplier:  Enzo Life Sciences ENZABS5600200
Encompass_Preferred
Clone: 735. Isotype: IgG2aκ. Immunogen: Homopolymers of α2,8-linked sialic acid of E. coli K1. Species reactivity: Human, Mouse, Rat. Specificity: Recognizes polysialic acid (PSA), a carbohydrate polymer attached to the neural cell adhesion molecule (NCAM), which shares molecular epitopes with homopolymers of α2,8-linked sialic acid of E. coli K1. Applications: Flow Cytometry, IHC, IP. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 0.02% Proclin 300.
Catalog No. 50-201-3086