Learn More
Cayman Chemical PolysarcosIn20 10mg
SDP
Catalog No. 502916178
Encompass_Preferred
An amine-functionalized hydrophilic polymer; addition to surface-grafted polymer brushes reduces protein absorption and mammalian and bacterial cell attachment
Catalog No. 50-291-6178 Supplier Cayman Chemical Supplier No. 3904810 MG
An amine-functionalized hydrophilic polymer; addition to surface-grafted polymer brushes reduces protein absorption and mammalian and bacterial cell attachment