Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Polysciences, Inc. Polysciences, Inc. Methacryloxyethyl thiocarbamoyl rhodamine B, Fluorescent Monomer, 100 mg
SDP

Catalog No. NC3991812
Encompass

This methacrylate-functionalized rhodamine B derivative is a fluorescent monomer designed for incorporation into polymer matrices and surface coatings. The thiocarbamoyl functional group enhances stability and reactivity in polymerization systems, allowing for varied fluorescence intensity by adjusting the amount of fluorescent monomer in the polymer.

  • Fluorescent monomer with excitation max: 548nm and emission max: 570nm.
  • Used in bioimaging, optical sensors, and labeling applications for high-sensitivity detection.
  • Integrated into temperature- and pH-responsive polymers for controlled fluorescence emission.
  • Enables covalent incorporation into methacrylate-based copolymers for functional materials.
  • Appearance: Purple crystals.
  • Handling requires gloves and a fume hood.
  • Storage at -20°C.

Catalog No. NC3991812 Supplier Polysciences, Inc. Supplier No. 23591100
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This methacrylate-functionalized rhodamine B derivative is a fluorescent monomer designed for incorporation into polymer matrices and surface coatings. The thiocarbamoyl functional group enhances stability and reactivity in polymerization systems, allowing for varied fluorescence intensity by adjusting the amount of fluorescent monomer in the polymer.

  • Fluorescent monomer with excitation max: 548nm and emission max: 570nm.
  • Used in bioimaging, optical sensors, and labeling applications for high-sensitivity detection.
  • Integrated into temperature- and pH-responsive polymers for controlled fluorescence emission.
  • Enables covalent incorporation into methacrylate-based copolymers for functional materials.
  • Appearance: Purple crystals.
  • Handling requires gloves and a fume hood.
  • Storage at -20°C.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.