Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Polyseed, Ricca Chemical
SDP

Catalog No. R586550050 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
50 Capsules
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R586550050 50 Capsules
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R586550050 Supplier Ricca Chemical Company Supplier No. R586550050
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Chemical Name or Material Polyseed
Physical Form Fine Crystals or Crystalline Powder
Quantity 50 Capsules
Grade Laboratory
Packaging Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.