Learn More
PolyShield™ Paraffin Repellent/Remover, StatLab™
GSA_VASDP
Catalog No. STLPS6
Change view
Click to view available options
Quantity:
6 x 120 mL
Catalog No. Quantity
STLPS6 6 x 120 mL
1 options
This item is not returnable. View return policy
Harmful if swallowed, if in contact with skin, or if inhaled.
Catalog No. STLPS6 Supplier SLMP Llc Dba Statlab Medical Supplier No. PS/6
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, if in contact with skin, or if inhaled.
PolyShield™ is a liquid paraffin repellent used as a protective coating on laboratory surfaces that are exposed to paraffin residue.
  • Can also be used to clean paraffin that has adhered to lab countertops and equipment
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Packaging Spray Bottle
Quantity 6 x 120 mL