Learn More
Electron Microscopy Sciences Polysine™ Adhesion Slide, 2x72/Bx
SDP
Catalog No. 5030436
Encompass_Preferred
The permanent bio-adhesive material on our Polysine slide attracts frozen and paraffin embedded sections, centrifuge preparation and cytology smears.
Catalog No. 50-304-36 Supplier Electron Microscopy Sciences Supplier No. 6341202
The permanent bio-adhesive material on our Polysine slide attracts frozen and paraffin embedded sections, centrifuge preparation and cytology smears.