Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Polysorbate 20, MP Biomedicals™

Catalog No. MP021947245 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP021947245 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP021947245 Supplier MP Biomedicals, Inc Supplier No. 02194724.5
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Polysorbate 20 is typically used as an emulsifying agent for the preparation of stable oil-in-water emulsions.

Tweens™ are a series of nonionic surfactants derived from sorbitan esters.

  • They are soluble or dispersible in water but differ widely in organic and oil solubilities.
  • Used as oil-in-water emulsifiers in pharmaceuticals, cosmetics, cleaning compounds, etc.
  • Polysorbate 20 is nonionic surfactants derived from sorbitan ester.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9005-64-5
Molecular Formula (C2H4O)y(C2H4O)w(C2H4O)x(C2H4O)zC18H34O6
Molecular Weight (g/mol) 522.68
MDL Number MFCD00165986
InChI Key HMFKFHLTUCJZJO-UHFFFAOYNA-N
Synonym tween 20, polysorbate 20, polyoxyethylene sorbitan monolaurate, polyoxyethylene 20 sorbitan monolaurate, polysorbate inn, 2-2-3,4-bis 2-hydroxyethoxy oxolan-2-yl-2-2-hydroxyethoxy ethoxy ethyl dodecanoate, alkest tw 20, polysorbate 20 nf, polysorbate 40 nf
PubChem CID 443314
IUPAC Name 2-[2-[3,4-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy]ethyl dodecanoate
SMILES CCCCCCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO

Specifications

Density 1.095g/mL
Quantity 500 mL
Physical Form Clear, Yellow to Amber Viscous Liquid
Refractive Index n20/D 1.468(lit.)
Specific Gravity 1.08–1.13
Viscosity 400 cps
Solubility Information Soluble/miscible in water to give a clear yellow solution; miscible with alcohol, dioxane, and ethyl acetate.
Formula Weight about 1,228g/mol
Grade Cell Culture Reagent
Concentration or Composition (by Analyte or Components) CriticalMicelle Concentration (CMC) 0.06 mM
Recommended Storage Store at Room Temperature (15–30°C)
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.