Learn More
Medchemexpress LLC Polysorbate 20, Polyoxyethylene(20) sorbitan monolaurate | 9005-64-5 | 98.0% | 100 ML
SDP
Catalog No. 5000364675
Encompass_Preferred
Tween 20 (Polysorbate 20) is a polyoxyethylene (POE)-type nonionic surfactant. It can modify lignin surface properties, altering hydrophobicity, hydrogen bonding, and surface charges to reduce non-productive adsorption of proteins. It has been shown to significantly increase the apparent solubility of drugs.
  • Nonionic surfactant
  • Modifies lignin surface properties
  • Reduces non-productive protein adsorption
  • Increases apparent drug solubility
  • For research use only
Catalog No. 50-003-64675 Supplier Medchemexpress LLC Supplier No. 100MLHY141415
Tween 20 (Polysorbate 20) is a polyoxyethylene (POE)-type nonionic surfactant. It can modify lignin surface properties, altering hydrophobicity, hydrogen bonding, and surface charges to reduce non-productive adsorption of proteins. It has been shown to significantly increase the apparent solubility of drugs.
  • Nonionic surfactant
  • Modifies lignin surface properties
  • Reduces non-productive protein adsorption
  • Increases apparent drug solubility
  • For research use only