Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Polysorbate 60, NF, Spectrum™ Chemical
SDP

Catalog No. 18605386 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
4 L
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-605-386 4 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-605-386 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. PO1364LTGL
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PO136, 9005-67-8

Spectrum™ Chemical Polysorbate 60, NF is used to solubilize essential oils. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 9005-67-8
Molecular Formula (C2H4O)y(C2H4O)w(C2H4O)x(C2H4O)zC24H46O6
Molecular Weight (g/mol) 522.68
MDL Number MFCD01780110
InChI Key DEKFZWMENCCOFU-UHFFFAOYNA-N
IUPAC Name 2-{2-[3,4-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl dodecanoate
SMILES CCCCCCCCCCCCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO

Specifications

Acid Value 2
CAS Max % 1
Quantity 4 L
Hydroxyl Value 81-96
Ignition Residue 0.0025
Packaging Amber Glass Bottle
Saponification Value 45-55
Water 0.03
Grade NF
Chemical Name or Material Polysorbate 60
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.