Learn More
Sigma Aldrich Fine Chemicals Biosciences Polysorbate 80, 9005-65-6, MFCD00082107, 100 G
Catalog No. NC0848536
Encompass
Tested according to Ph Eur, Synonym: Polyoxyethylenesorbitan monooleate, Polysorbatum 80, TWEEN 80. Polysorbate 80 has been used in drug (rapamycin) treatment and to study the effect of rapamycin on osteolysis and tumor growth. It has also been used in the preparation of spray-dried emulsions and to study antibacterial efficacy.
Catalog No. NC0848536 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 59924100GF
Tested according to Ph Eur, Synonym: Polyoxyethylenesorbitan monooleate, Polysorbatum 80, TWEEN 80. Polysorbate 80 has been used in drug (rapamycin) treatment and to study the effect of rapamycin on osteolysis and tumor growth. It has also been used in the preparation of spray-dried emulsions and to study antibacterial efficacy.