Learn More
Andwin Scientific POLYSORBATE 80 BDH SOL 500ML
SDP
Catalog No. NC0518204
Encompass_Preferred
The Polysorbate solution is a viscous clear, colorless liquid chemical that is composed of 100% Polyoxyethylene (80) Sorbitan Monooleate. This product is packaged and sold in a 500 mL amber glass bottle.
Catalog No. NC0518204 Supplier Andwin Scientific Supplier No. BDH77812
The Polysorbate solution is a viscous clear, colorless liquid chemical that is composed of 100% Polyoxyethylene (80) Sorbitan Monooleate. This product is packaged and sold in a 500 mL amber glass bottle.