Learn More
Sigma Aldrich Fine Chemicals Biosciences Polysorbate 80 fG25KG
Catalog No. 502902103
Encompass
Polysorbate 80 (TWEEN(R) 80) is a polyethylene sorbitol ester. It has been tested as an efficient phase transfer catalyst for cyanation of alkyl halides Williamson synthesis of ether and base-catalyzed dehydrobromination of alkyl halides.
Catalog No. 50-290-2103 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. W29170625KGK
Polysorbate 80 (TWEEN(R) 80) is a polyethylene sorbitol ester. It has been tested as an efficient phase transfer catalyst for cyanation of alkyl halides Williamson synthesis of ether and base-catalyzed dehydrobromination of alkyl halides.