Learn More
LAB ALLEY Lab Alley Polysorbate 80 Food Grade Kosher Halal
SDP
Supplier:  LAB ALLEY PS80F1GAL
Encompass
Polysorbate 80 is a nonionic surfactant and emulsifier often used in foods and cosmetics. This synthetic compound is a viscous water-soluble yellow liquid. Polysorbate 80 is a hydrophilic nonionic surfactant. It is utilized as a surfactant in soaps and cosmetics and also as a lubricant in eye drops. In food or pharmaceutical products it can act as an emulsifier. Polysorbate 80 is used as an emulsifier or defoamer in foods vitamins medicines and vaccines. 
Catalog No. NC2945444