Learn More
Sigma Aldrich Fine Chemicals Biosciences Polysorbate 80 tested acco1KG
Catalog No. 502899452
Encompass
Polysorbate 80 is a surfactant. Polysorbates are a combination of oleic esters of sorbitol and sorbitanes with polyethylene glycol chains.
Catalog No. 50-289-9452 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 599241KGF
Polysorbate 80 is a surfactant. Polysorbates are a combination of oleic esters of sorbitol and sorbitanes with polyethylene glycol chains.