Learn More
Crescent Chemical Co Inc POLYSORBATE 80 VG
SDP
Catalog No. 50248026
Encompass_Preferred
Polysorbate 80 VG The fatty acids of this detergent are of vegetable origin. Size - 500G Storage Conditions - +15 °C TO +30 °C Catalog Number - 33116.01 CAS 9005-65-6
Catalog No. 50-248-026 Supplier Crescent Chemical Co Inc Supplier No. SE33116.01
Polysorbate 80 VG The fatty acids of this detergent are of vegetable origin. Size - 500G Storage Conditions - +15 °C TO +30 °C Catalog Number - 33116.01 CAS 9005-65-6