Learn More
Med Vet International First Aid Antibiotic Polysporin Ointment 0.9 Gram Individual Packet, 10312547238134, 144/Box
Catalog No. 503044469
Encompass_Preferred
Dual antibiotic combines the power of bacitracin with polymyxin B sulfate 1 gram contains bacitracin 500 units; polymyxin b 10,000 units Does not contain neomycin so it's safe for those who will experience negative side effects if they use Neosporin 0.9 gram per packet 144 packets makes this box a good choice for a workplace first-aid kit or your travel toiletries bag "just in case"
Catalog No. 50-304-4469 Supplier Med Vet International Supplier No. 565780BX
Dual antibiotic combines the power of bacitracin with polymyxin B sulfate 1 gram contains bacitracin 500 units; polymyxin b 10,000 units Does not contain neomycin so it's safe for those who will experience negative side effects if they use Neosporin 0.9 gram per packet 144 packets makes this box a good choice for a workplace first-aid kit or your travel toiletries bag "just in case"