Learn More
Med Vet International First Aid Antibiotic Polysporin Ointment 1 oz. Tube, 00300810237895, Each
Catalog No. 503044656
Encompass_Preferred
Dual antibiotic combines the power of bacitracin with polymyxin B 1 gram contains 500 units of bacitracin and 10,000 units polymyxin B Does not contain neomycin so it's safe for those who will experience negative side effects when they use Neosporin 28.3 gram tube
Catalog No. 50-304-4656 Supplier Med Vet International Supplier No. 495868EA
Dual antibiotic combines the power of bacitracin with polymyxin B 1 gram contains 500 units of bacitracin and 10,000 units polymyxin B Does not contain neomycin so it's safe for those who will experience negative side effects when they use Neosporin 28.3 gram tube