Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Streamlight™ PolyStinger™ Flashlight

Catalog No. 19822944 Shop All Streamlight Products
Change view
Click to view available options
Color:
Black
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-822-944 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-822-944 Supplier Streamlight™ Supplier No. 76521

missing translation for 'validMainframeMsg'

Includes

Spare bulb in tailcap and AC/DC chargers and holders

Suitable for use in marine, damp, and corrosive environments

  • Powerful, rechargeable flashlight with Xenon bi-pin bulb provides up to 11,000 candela with 90 minutes of continuous run time between charges
  • Recharges on 110VAC or 12VDC steady chargers with 10-hour charge time using
  • Fast charger enables 2.5 hour charge time
  • Constructed of yellow, super-tough, nonconductive polymer with rubber nonslip grip
  • Compact snap-in charger recharges NiCd battery up to 1000 times; LED light indicates when flashlight is fully charged

Order Info

Length: 7.34 in. (18.7cm). Weight: 8.7 oz. (247g).

Certifications

UL listed for certain Class I, Division 2 hazardous environments; CE marked.

Specifications

Gas Fill Xenon
Voltage 110 V
Weight (English) 8.7 oz.
Weight (Metric) 0.847 kg
Battery Type NiCad
Color Black
Controller Type Push-button head switch
Diameter (English) Body 1.15 in.
Diameter (Metric) Body 2.92 cm
For Use With (Application) For use in marine, damp, and corrosive environments
Includes Spare bulb in tail cap and AC/DC chargers and holders
Length (English) 7.34 in.
Length (Metric) 18.4 cm
Material Nylon Polymer
Product Type Rechargeable Flashlight
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.