Learn More
Ted Pella Inc 6-Compartment Styrene Box
SDP
Catalog No. NC1557510
Encompass
Clear Polystyrene Plastic Sorting Boxes, with Hinged Cover. 6 compartments: 4-3/16" x 3-1/2" x 1/2" (10.64 x 8.89 x 1.27cm). inside size compartments: 1-1/3" x 1-2/3" x 3/8" (3.4 x 4.2 x 1.0cm)
Catalog No. NC1557510 Supplier Ted Pella Inc Supplier No. 139201
Clear Polystyrene Plastic Sorting Boxes, with Hinged Cover. 6 compartments: 4-3/16" x 3-1/2" x 1/2" (10.64 x 8.89 x 1.27cm). inside size compartments: 1-1/3" x 1-2/3" x 3/8" (3.4 x 4.2 x 1.0cm)