Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Polystyrene, Analytical Standard, For GPC, 20,000, MilliporeSigma™ Supelco™

Catalog No. 111004721 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-100-4721 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-100-4721 Supplier MilliporeSigma Supelco Supplier No. 3277431G
Add to Cart
Add to Cart

Polystyrene (for GPC, 20,000) is a synthetic thermoplastic, that finds a variety of applications owing to its characteristics like low cost, rigidity, low specific weight, high chemical resistance, mechanical flexibility, biocompatibility and good processability.

TRUSTED_SUSTAINABILITY

Specifications

Packaging Glass Bottle
Physical Form Powder
Linear Formula [CH2CH(C6H5)]n
MDL Number MFCD00084450
Synonym PS
Quantity 1 g
Grade GPC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.